Learn More
2-Methoxybenzenemethanol, TRC
Product Code. 30470252
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity Unit Size
30470252 25 g 25g
1 options
This item is not returnable. View return policy
Product Code. 30470252 Supplier Toronto Research Chemicals Supplier No. TRCM26076025G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methoxybenzenemethanol
CAS 612-16-8
Molecular Formula C8H10O2
Quantity 25 g
Synonym (2-Methoxyphenyl)methanol, 2-(Hydroxymethyl)anisole, 2-Methoxybenzenemethanol, 2-Methoxybenzyl alcohol, NSC 66558, o-Anisalcohol, o-Methoxybenzyl Alcohol, o-Methoxyphenylmethanol
InChI Formula InChI=1S/C8H10O2/c1-10-8-5-3-2-4-7(8)6-9/h2-5,9H,6H2,1H3
SMILES COc1ccccc1CO
IUPAC Name (2-methoxyphenyl)methanol
Molecular Weight (g/mol) 138.16
Formula Weight 138.0681