missing translation for 'onlineSavingsMsg'
Learn More

Ácido 2-metoxi-5-sulfamoilbenzoico, 97 %, Thermo Scientific Chemicals

Catalog Number p-7027158
Click to view available options
Cantidad:
10 g
50 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
22117-85-7
C8H9NO5S
231.222
MFCD00129997
SQAILWDRVDGLGY-UHFFFAOYSA-N
2-methoxy-5-aminosulfonylbenzoic acid, benzoic acid, 5-aminosulfonyl-2-methoxy, 5-sulphamoyl-o-anisic acid, 2-methoxy-5-sulfonylbenzoic acid, sulpiride impurity d, acmc-209fsc, 2-methoxy-5-sulfamoylbenzoicacid, 2-methoxy-5-sulphamoylbenzoic acid, 2-methoxy-5-sulfamoyl benzoic acid, 2-methoxy-5-sulfamoyl-benzoic acid
89601
ácido 2-metoxi-5-sulfamoilbenzoico
COC1=C(C=C(C=C1)S(=O)(=O)N)C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
22117-85-7
C8H9NO5S
231.222
MFCD00129997
SQAILWDRVDGLGY-UHFFFAOYSA-N
2-methoxy-5-aminosulfonylbenzoic acid, benzoic acid, 5-aminosulfonyl-2-methoxy, 5-sulphamoyl-o-anisic acid, 2-methoxy-5-sulfonylbenzoic acid, sulpiride impurity d, acmc-209fsc, 2-methoxy-5-sulfamoylbenzoicacid, 2-methoxy-5-sulphamoylbenzoic acid, 2-methoxy-5-sulfamoyl benzoic acid, 2-methoxy-5-sulfamoyl-benzoic acid
89601
ácido 2-metoxi-5-sulfamoilbenzoico
COC1=C(C=C(C=C1)S(=O)(=O)N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Methoxy-5-sulfamoylbenzoic acid
Cantidad 10 g
Formula Weight (peso de la fórmula) 232.23
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'