Learn More
Hirschmann™ 2 m Opus™ Data Power Cable
Product Code. 10217873
Change view
Click to view available options
Length (Metric):
2 m
Unit Size:
Each
Product Code. Length (Metric) Unit Size
10217873 2 m Each
1 options
This item is not returnable. View return policy
Product Code. 10217873 Supplier Hirschmann™ Supplier No. 9587002
  • ONLINE EXCLUSIVE PRICE
    174.30€
    114.71€ / paquete de 1
    Ahorro 59.59€
    34% Descuento
Fecha estimada de envoi: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A connection cable for Opus dispensers and the control unit.
  • 2 m in length
  • Connects basic unit to control unit
  • Accessory item
Specifications
Length (Metric) 2 m
For Use With (Equipment) Opus Dispensers
For Use With (Application) Connecting Basic Unit to Control Unit
Type Data Power Cable