missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%

Product Code. 11360164
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360164 25 g 25g
11370164 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360164 Supplier Thermo Scientific Alfa Aesar Supplier No. L00679.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7237-34-5 | C20H20BrOP | 387.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7237-34-5
Molecular Formula C20H20BrOP
Molecular Weight (g/mol) 387.26
MDL Number MFCD00011901
InChI Key QZJOQNHOOVSESC-UHFFFAOYSA-M
Synonym 2-hydroxyethyl triphenylphosphonium bromide, 2-hydroxyethyl triphenylphosphanium bromide, acmc-20appe, 2-hydroxyethyltriphenylphosphonium bromide, 2-hydroxyethyl triphenyphosphonium bromide, 2-hydroxyethyl triphenyl phosphanium bromide, 2-hydroxy ethyl triphenylphosphonium bromide, phosphonium, 2-hydroxyethyl triphenyl-,bromide 1:1, phosphonium, 2-hydroxyethyl triphenyl-, bromide 1:1
PubChem CID 2733550
IUPAC Name 2-hydroxyethyl(triphenyl)phosphanium;bromide
SMILES [Br-].OCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 218°C to 220°C
Quantity 25 g
Beilstein 3922741
Sensitivity Hygroscopic
Formula Weight 387.26
Percent Purity ≥98%
Chemical Name or Material (2-Hydroxyethyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.