Learn More
2-Hydroxy-5-(trifluoromethyl)pyridine, TRC
Product Code. 30240918
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240918 100 g 100g
30240822 5 g 5g
30240571 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240918 Supplier Toronto Research Chemicals Supplier No. TRCH971570100G
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Añadir a la cesta
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
CAS 33252-63-0
Molecular Formula C6 H4 F3 N O
Molecular Weight (g/mol) 163.1
Synonym 5-(Trifluoromethyl)-2-pyridinol, 2-Pyridinol, 5-(trifluoromethyl)-
IUPAC Name 5-(trifluoromethyl)pyridin-2-ol
SMILES Oc1ccc(cn1)C(F)(F)F
Especificaciones
Quantity 100 g
InChI Formula InChI=1S/C6H4F3NO/c7-6(8,9)4-1-2-5(11)10-3-4/h1-3H,(H,10,11)
Formula Weight 163.0245
Chemical Name or Material 2-Hydroxy-5-(trifluoromethyl)pyridine