missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorofenilurea, 98 %, Thermo Scientific Chemicals

Catalog Number p-7043548
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
656-31-5
C7H7FN2O
154.144
MFCD00014786
PAWVOCWEWJXILY-UHFFFAOYSA-N
2-fluorophenyl urea, 1-2-fluorophenyl urea, o-fluorophenylurea, n-2-fluorophenyl urea, amino-n-2-fluorophenyl amide, o-fluorophenyl urea, acmc-20anu9, urea, o-fluorophenyl, urea,n-2-fluorophenyl
12606
(2-fluorofenil)urea
C1=CC=C(C(=C1)NC(=O)N)F
Este artículo no se puede devolver. Vea la política de devoluciones
656-31-5
C7H7FN2O
154.144
MFCD00014786
PAWVOCWEWJXILY-UHFFFAOYSA-N
2-fluorophenyl urea, 1-2-fluorophenyl urea, o-fluorophenylurea, n-2-fluorophenyl urea, amino-n-2-fluorophenyl amide, o-fluorophenyl urea, acmc-20anu9, urea, o-fluorophenyl, urea,n-2-fluorophenyl
12606
(2-fluorofenil)urea
C1=CC=C(C(=C1)NC(=O)N)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Fluorophenylurea
Punto de fusión 178°C to 184°C
Cantidad 5 g
Beilstein 2803651
Formula Weight (peso de la fórmula) 154.14
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'