Learn More
2-Dodecanona, 98 %, Thermo Scientific Chemicals
Catalog Number p-7024353
Click to view available options
Cantidad:
10 g
50 g
6175-49-1
C12H24O
184.323
MFCD00015064
LSKONYYRONEBKA-UHFFFAOYSA-N
2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc
22556
Dodecan-2-ona
CCCCCCCCCCC(=O)C
6175-49-1
C12H24O
184.323
MFCD00015064
LSKONYYRONEBKA-UHFFFAOYSA-N
2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc
22556
Dodecan-2-ona
CCCCCCCCCCC(=O)C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Dodecanone
Punto de fusión 17°C to 20°C
Densidad 0.82
Punto de ebullición 247°C
Temperatura de inflamación 110°C (230°F)
Índice de refracción 1.433
Cantidad 10 g
Beilstein 1703135
Formula Weight (peso de la fórmula) 184.32
Porcentaje de pureza ≥98%