Learn More
2-(Difluoromethoxy)benzaldehyde, 97%, Thermo Scientific™
Product Code. 11399839
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399839 5 g 5g
11389839 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399839 Supplier Thermo Scientific Alfa Aesar Supplier No. B24272.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-(Difluoromethoxy)benzaldehyde is an important raw material used as an intermediate in the preparation of pharmaceuticals, agrochemicals, and dyestuffs. It is also involved in the preparation of 4-(2-difluoromethoxyphenyl)- substituted 1,4-dihydropyridine 3,5-diesters.
Solubility
Miscible with 2-propanol and acetic acid.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71653-64-0
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042251
InChI Key QPBNHDFPMRENBC-UHFFFAOYSA-N
Synonym 2-difluoromethoxy benzaldehyde, 2-difluoromethoxy-benzaldehyde, o-difluoromethoxybenzaldehyde, 2-difloromethoxybenzaldehyde, pubchem2906, acmc-209ojk, difluoromethoxybenzaldehyde, ksc356m9l, zerenex zx009675, benzaldehyde, difluoromethoxy
PubChem CID 2736984
IUPAC Name 2-(difluoromethoxy)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)OC(F)F
Specifications
Density 1.284
Boiling Point 88°C to 90°C (9 mmHg)
Flash Point 106°C (223°F)
Refractive Index 1.497
Quantity 5 g
Beilstein 389068
Sensitivity Air Sensitive
Solubility Information Miscible with 2-propanol and acetic acid.
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)benzaldehyde