Learn More
2-(Diethylamino)ethyl 4-Amino-3-hydroxybenzoate Hydrochloride, Mikromol™
Product Code. 30050269
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30050269 25 mg 25mg
1 options
This item is not returnable. View return policy
Product Code. 30050269 Supplier Mikromol Supplier No. MM0543.070025
 Restricted Item
Fecha estimada de envoi: 19-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(Diethylamino)ethyl 4-Amino-3-hydroxybenzoate Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(Diethylamino)ethyl 4-Amino-3-hydroxybenzoate Hydrochloride
CAS 860693-48-7
Analyte or Component Names 2-(Diethylamino)ethyl 4-Amino-3-hydroxybenzoate Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H20 N2 O3 . Cl H
Certifications/Compliance ISO 9001
Synonym 4-Amino-3-hydroxybenzoic Acid 2-(Diethylamino)ethyl Ester Hydrochloride (1:1)
InChI Formula InChI=1S/C13H20N2O3.ClH/c1-3-15(4-2)7-8-18-13(17)10-5-6-11(14)12(16)9-10;/h5-6,9,16H,3-4,7-8,14H2,1-2H3;1H
SMILES Cl.CCN(CC)CCOC(=O)c1ccc(N)c(O)c1
IUPAC Name 2-(diethylamino)ethyl 4-amino-3-hydroxybenzoate;hydrochloride
Molecular Weight (g/mol) 288.77
Quantity 25 mg
Formula Weight 288.1241 g/mol