missing translation for 'onlineSavingsMsg'
Learn More

Fosforodicloridato de 2-clorofenilo, + 98 %, Thermo Scientific Chemicals

Catalog Number p-7081502
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
15074-54-1
C6H4Cl3O2P
245.42
MFCD00009704
VLDPXPPHXDGHEW-UHFFFAOYSA-N
2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
84775
1-cloro-2-diclorofosforiloxibenceno
C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
15074-54-1
C6H4Cl3O2P
245.42
MFCD00009704
VLDPXPPHXDGHEW-UHFFFAOYSA-N
2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
84775
1-cloro-2-diclorofosforiloxibenceno
C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chlorophenyl phosphorodichloridate
Densidad 1.491
Punto de ebullición 82°C to 84°C (0.4 mmHg)
Temperatura de inflamación 105°C (221°F)
Índice de refracción 1.538
Cantidad 5 g
Número UN UN3265
Beilstein 911009
Sensibilidad Moisture sensitive
Formula Weight (peso de la fórmula) 245.43
Porcentaje de pureza ≥98%
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'