Learn More
Metacrilato de 2-cloroetilo, 97 %, estabilizado con 500 ppm de hidroquinona, Thermo Scientific Chemicals
Catalog Number p-7045200
Click to view available options
Cantidad:
10 g
50 g
1888-94-4
C6H9ClO2
148.59
MFCD00045291
GPOGMJLHWQHEGF-UHFFFAOYSA-N
2-chloroethyl methacrylate, chloroethyl methacrylate, methacrylic acid, 2-chloroethyl ester, 2-propenoic acid, 2-methyl-, 2-chloroethyl ester, .beta.-chloroethyl methacrylate, methacrylic acid beta-chloroethyl ester, methacrylic acid .beta.-chloroethyl ester, acmc-20anxx, chloroethylmethacrylate, 2-chloroethylmethacrylate
15905
2-metilprop-2-enoato de 2-cloroetilo
CC(=C)C(=O)OCCCl
1888-94-4
C6H9ClO2
148.59
MFCD00045291
GPOGMJLHWQHEGF-UHFFFAOYSA-N
2-chloroethyl methacrylate, chloroethyl methacrylate, methacrylic acid, 2-chloroethyl ester, 2-propenoic acid, 2-methyl-, 2-chloroethyl ester, .beta.-chloroethyl methacrylate, methacrylic acid beta-chloroethyl ester, methacrylic acid .beta.-chloroethyl ester, acmc-20anxx, chloroethylmethacrylate, 2-chloroethylmethacrylate
15905
2-metilprop-2-enoato de 2-cloroetilo
CC(=C)C(=O)OCCCl
2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El metacrilato de 2-cloroetilo se utiliza como monómero para preparar poli(metacrilato de 2-cloroetilo).
Solubilidad
No miscible con agua.
Notas
Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloroethyl methacrylate
Nombre de nota stabilized with 500ppm hydroquinone
Punto de ebullición 170°C
Temperatura de inflamación 69°C (156°F)
Índice de refracción 1.451
Cantidad 10 g
Número UN UN2810
Beilstein 956599
Información de solubilidad Immiscible with water.
Formula Weight (peso de la fórmula) 148.59
Porcentaje de pureza 97%