Learn More
2-Chloro-N-[(furan-2-yl)methyl]-7H-purin-6-amine, TRC
Product Code. 30363408
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363408 100 mg 100mg
30363130 500 mg 500mg
30354837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363408 Supplier Toronto Research Chemicals Supplier No. TRCC369853100MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101862-47-9
Molecular Formula C10H8N5OCl
Molecular Weight (g/mol) 249.66
Synonym 2-Chloro-N-(2-furanylmethyl)-9H-purin-6-amine, (2-Chloro-7H-purin-6-yl)[(furan-2-yl)methyl]amine, 2-Chloro-6-Furfurylaminopurine, 2-Chloro-N-(2-furanylmethyl)-7H-purin-6-amine, NSC 45794
IUPAC Name 2-chloro-N-(furan-2-ylmethyl)-7H-purin-6-amine
SMILES Clc1nc(NCc2occc2)c3ncnc3[nH]1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H8ClN5O/c11-10-15-8(7-9(16-10)14-5-13-7)12-4-6-2-1-3-17-6/h1-3,5H,4H2,(H2,12,13,14,15,16)
Formula Weight 249.0417
Chemical Name or Material 2-Chloro-N-[(furan-2-yl)methyl]-7H-purin-6-amine