missing translation for 'onlineSavingsMsg'
Learn More

2-Cloro-5-(trifluorometil)fenilacetonitrilo, 97 %

Catalog Number p-7026997
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
22902-88-1
C9H5ClF3N
219.59
MFCD01631472
KTMVZYQQKJZSFK-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl phenylacetonitrile, 2-chloro-5-trifluoromethylbenzylcyanide, 2-chloro-5-trifluoromethylbenzyl cyanide, 2-2-chloro-5-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile,2-chloro-5-trifluoromethyl, 2-2-chloro-5-trifluoromethyl phenyl ethanenitrile, acmc-20aicj, 4-chloro-3-cyanomethyl benzotrifluoride, 2-chloro-5-trifluoromethyl-phenyl-acetonitrile, 2-chloro-5-trifluoromethyl phenyl acetonitrile
2778124
2-[2-cloro-5-(trifluorometil)fenil]acetonitrilo
FC(F)(F)C1=CC=C(Cl)C(CC#N)=C1
Este artículo no se puede devolver. Vea la política de devoluciones
22902-88-1
C9H5ClF3N
219.59
MFCD01631472
KTMVZYQQKJZSFK-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl phenylacetonitrile, 2-chloro-5-trifluoromethylbenzylcyanide, 2-chloro-5-trifluoromethylbenzyl cyanide, 2-2-chloro-5-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile,2-chloro-5-trifluoromethyl, 2-2-chloro-5-trifluoromethyl phenyl ethanenitrile, acmc-20aicj, 4-chloro-3-cyanomethyl benzotrifluoride, 2-chloro-5-trifluoromethyl-phenyl-acetonitrile, 2-chloro-5-trifluoromethyl phenyl acetonitrile
2778124
2-[2-cloro-5-(trifluorometil)fenil]acetonitrilo
FC(F)(F)C1=CC=C(Cl)C(CC#N)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloro-5-(trifluoromethyl)phenylacetonitrile
Punto de fusión 30°C to 34°C
Punto de ebullición 140°C to 142°C (12 mmHg)
Cantidad 1 g
Número UN UN3439
Beilstein 1960651
Formula Weight (peso de la fórmula) 219.59
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'