missing translation for 'onlineSavingsMsg'
Learn More

2-Cloro-5-nitroanilina, 98 %, Thermo Scientific Chemicals

Catalog Number p-7022365
Click to view available options
Cantidad:
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
6283-25-6
C6H5ClN2O2
172.568
MFCD00007668
KWIXNFOTNVKIGM-UHFFFAOYSA-N
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
22691
2-cloro-5-nitroanilina
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
6283-25-6
C6H5ClN2O2
172.568
MFCD00007668
KWIXNFOTNVKIGM-UHFFFAOYSA-N
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
22691
2-cloro-5-nitroanilina
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloro-5-nitroaniline
Punto de fusión 117°C to 121°C
Temperatura de inflamación 191°C (375°F)
Olor Weak, Characteristic
Cantidad 100 g
Número UN UN2237
Beilstein 2208878
Formula Weight (peso de la fórmula) 172.57
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'