Learn More
Ácido 2-cloro-4-fluoro-3-metilbenzoico, 97 %, Thermo Scientific Chemicals
Catalog Number p-7034212
Click to view available options
Cantidad:
250 mg
1 g
173315-54-3
C8H6ClFO2
188.58
MFCD11846444
DBMUFFHLBXKTDM-UHFFFAOYSA-N
benzoic acid, 2-chloro-4-fluoro-3-methyl, 2-chloro-4-fluoro-3-methyl-benzoic acid, 2-chloro-3-methyl-4-fluoro-benzoic acid, 2-chloro-4-fluoro-3-methyl benzoic acid
10845315
ácido 2-cloro-4-fluoro-3-metilbenzoico
CC1=C(F)C=CC(C(O)=O)=C1Cl
173315-54-3
C8H6ClFO2
188.58
MFCD11846444
DBMUFFHLBXKTDM-UHFFFAOYSA-N
benzoic acid, 2-chloro-4-fluoro-3-methyl, 2-chloro-4-fluoro-3-methyl-benzoic acid, 2-chloro-3-methyl-4-fluoro-benzoic acid, 2-chloro-4-fluoro-3-methyl benzoic acid
10845315
ácido 2-cloro-4-fluoro-3-metilbenzoico
CC1=C(F)C=CC(C(O)=O)=C1Cl
It is used in the synthesis and Antibacterial Evaluation of 6-Amino-8-methylquinolones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Se utiliza en la síntesis y evaluación antibacteriana de 6-amino-8-metilquinolonas.
Notas
Almacenar alejado de agentes oxidantes. Mantener el recipiente bien cerrado en un lugar fresco, seco y bien ventilado.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloro-4-fluoro-3-methylbenzoic acid
Punto de fusión 186°C to 189°C
Cantidad 250 mg
Beilstein 7423351
Formula Weight (peso de la fórmula) 188.59
Porcentaje de pureza 97%