missing translation for 'onlineSavingsMsg'
Learn More

Ácido 2-cloro-4-(1-pirrolidinilcarbonil)bencenoborónico, 97 %, Thermo Scientific™

Catalog Number p-7036867
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1449131-93-4
C11H13BClNO3
253.489
MFCD22125137
CCPHBMWIZPSWRQ-UHFFFAOYSA-N
2-chloro-4-1-pyrrolidinylcarbonyl phenylboronic acid, 2-chloro-4-1-pyrrolidinylcarbonyl benzeneboronic acid, 2-chloro-4-pyrrolidine-1-carbonyl phenylboronic acid, 2-chloro-4-pyrrolidine-1-carbonyl phenyl boronic acid
73996117
ácido [2-cloro-4-(pirrolidin-1-carbonil)fenil]borónico
B(C1=C(C=C(C=C1)C(=O)N2CCCC2)Cl)(O)O
Este artículo no se puede devolver. Vea la política de devoluciones
1449131-93-4
C11H13BClNO3
253.489
MFCD22125137
CCPHBMWIZPSWRQ-UHFFFAOYSA-N
2-chloro-4-1-pyrrolidinylcarbonyl phenylboronic acid, 2-chloro-4-1-pyrrolidinylcarbonyl benzeneboronic acid, 2-chloro-4-pyrrolidine-1-carbonyl phenylboronic acid, 2-chloro-4-pyrrolidine-1-carbonyl phenyl boronic acid
73996117
ácido [2-cloro-4-(pirrolidin-1-carbonil)fenil]borónico
B(C1=C(C=C(C=C1)C(=O)N2CCCC2)Cl)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloro-4-(1-pyrrolidinylcarbonyl)benzeneboronic acid
Cantidad 250 mg
Formula Weight (peso de la fórmula) 253.49
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'