missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cloro-2'-arabino-fluoro-2'-desoxiadenosina, + 99 %, Thermo Scientific Chemicals
Click to view available options
Cantidad:
50 mg
100 mg
250 mg
Descripción
A purine nucleoside anti-metabolite. Toxic to nondividing lymphocytes and monocytes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Un antimetabolito del nucleósido purínico
- Tóxico para los linfocitos y monocitos no divisibles
Especificaciones
Nombre del producto químico o material | 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine |
Color | Blanco |
Cantidad | 50 mg |
Número UN | UN2811 |
Índice Merck | 14,2372 |
Formula Weight (peso de la fórmula) | 303.68 |
Porcentaje de pureza | ≥99% |
Forma física | Polvo |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'