Learn More
2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde, Mikromol™
Description
2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde
Specifications
Specifications
| Chemical Name or Material | 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde |
| CAS | 83857-96-9 |
| Analyte or Component Names | 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H11 Cl N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Losartan Potassium Imp. D (EP), Losartan Imp. D (EP), Losartan USP Related Compound A, Losartan USP RC A, 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde, Losartan Related Compound A, Losartan Potassium Impurity D, Losartan Impurity D |
| InChI Formula | InChI=1S/C8H11ClN2O/c1-2-3-4-7-10-6(5-12)8(9)11-7/h5H,2-4H2,1H3,(H,10,11) |
| SMILES | CCCCc1nc(Cl)c(C=O)[nH]1 |
| IUPAC Name | 2-butyl-4-chloro-1H-imidazole-5-carbaldehyde |
| Show More |