missing translation for 'onlineSavingsMsg'
Learn More

2-Buteno-1,4-bis(cloruro de trifenilfosfonio), + 98 %

Catalog Number p-7043384
Click to view available options
Cantidad:
10 g
50 g
Este artículo no se puede devolver. Vea la política de devoluciones
106423-29-4
C40H36Cl2P2
649.576
MFCD00031713
OOACLPDZHDFCQP-UHFFFAOYSA-L
2-butene-1,4-bis triphenylphosphonium chloride, trans-2-butene-1,4-bis triphenylphosphoniumchloride, but-2-ene-1,4-diyl bis triphenylphosphanium dichloride
70700154
trifenil(4-trifenilfosmanioilbut-2-enil)fosferanio; dicloruro
C1=CC=C(C=C1)[P+](CC=CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Cl-].[Cl-]
Este artículo no se puede devolver. Vea la política de devoluciones
106423-29-4
C40H36Cl2P2
649.576
MFCD00031713
OOACLPDZHDFCQP-UHFFFAOYSA-L
2-butene-1,4-bis triphenylphosphonium chloride, trans-2-butene-1,4-bis triphenylphosphoniumchloride, but-2-ene-1,4-diyl bis triphenylphosphanium dichloride
70700154
trifenil(4-trifenilfosmanioilbut-2-enil)fosferanio; dicloruro
C1=CC=C(C=C1)[P+](CC=CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Cl-].[Cl-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Butene-1,4-bis(triphenylphosphonium chloride)
Punto de fusión 264°C to 267°C
Cantidad 10 g
Beilstein 3866753
Formula Weight (peso de la fórmula) 649.6
Porcentaje de pureza ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'