Learn More
2-Bromoindeno, 98 %, Thermo Scientific Chemicals
Catalog Number p-7028257
Click to view available options
Cantidad:
1 g
5 g
25 g
10485-09-3
C9H7Br
195.059
MFCD06797863
CCUYEVNCRQDQRF-UHFFFAOYSA-N
2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate
575586
2-bromo-1H-indeno
C1C2=CC=CC=C2C=C1Br
10485-09-3
C9H7Br
195.059
MFCD06797863
CCUYEVNCRQDQRF-UHFFFAOYSA-N
2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate
575586
2-bromo-1H-indeno
C1C2=CC=CC=C2C=C1Br
2-Bromoindene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El 2-bromoindeno se utiliza como producto intermedio farmacéutico.
Solubilidad
Insoluble en agua.
Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Bromoindene
Punto de fusión 35°C to 37°C
Cantidad 1 g
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 195.06
Porcentaje de pureza 98%