Learn More
2-Bromoacetylamino-3-(2-chlorobenzoyl)-5-(2-carbomethoxyethyl)thiophene, TRC
Product Code. 30343597
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
30343597 5 mg 5mg
1 options
This item is not returnable. View return policy
Product Code. 30343597 Supplier Toronto Research Chemicals Supplier No. TRCB6790855MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Bromoacetylamino-3-(2-chlorobenzoyl)-5-(2-carbomethoxyethyl)thiophene
CAS 100827-79-0
Molecular Formula C17H15BrClNO4S
Quantity 5 mg
Synonym 5-[(2-Bromoacetyl)amino]-4-(2-chlorobenzoyl)-2-thiophenepropanoic Acid Methyl Ester, 5-[(Bromoacetyl)amino]-4-(2-chlorobenzoyl)-2-thiophenepropanoic Acid Methyl Ester
InChI Formula InChI=1S/C17H15BrClNO4S/c1-24-15(22)7-6-10-8-12(17(25-10)20-14(21)9-18)16(23)11-4-2-3-5-13(11)19/h2-5,8H,6-7,9H2,1H3,(H,20,21)
SMILES COC(=O)CCc1cc(C(=O)c2ccccc2Cl)c(NC(=O)CBr)s1
IUPAC Name methyl 3-[5-[(2-bromoacetyl)amino]-4-(2-chlorobenzoyl)thiophen-2-yl]propanoate
Molecular Weight (g/mol) 444.73