Learn More
2-Bromo-6-piridinametanol, 96 %, Thermo Scientific Chemicals
Catalog Number p-7037400
Click to view available options
Cantidad:
250 mg
1 g
5 g
33674-96-3
C6H6BrNO
188.02
MFCD04039884
XDDGKNRSCDEWBR-UHFFFAOYSA-N
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
3517811
(6-bromopiridin-2-il)metanol
OCC1=CC=CC(Br)=N1
33674-96-3
C6H6BrNO
188.02
MFCD04039884
XDDGKNRSCDEWBR-UHFFFAOYSA-N
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
3517811
(6-bromopiridin-2-il)metanol
OCC1=CC=CC(Br)=N1
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Es un principio farmacéutico activo y un adyuvante farmacéutico.
Solubilidad
Ligeramente soluble en agua.
Notas
Incompatible con agentes oxidantes y bases. Almacenar a temperatura ambiente.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Bromo-6-pyridinemethanol
Punto de fusión 34°C to 39°C
Punto de ebullición 246°C
Temperatura de inflamación >110°C (230°F)
Cantidad 250 mg
Información de solubilidad Slightly soluble in water.
Formula Weight (peso de la fórmula) 188.02
Porcentaje de pureza 96%