missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de 2-bromo-6-fluorobencilamina, 96 %, Thermo Scientific Chemicals

Catalog Number 15457006
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
1214330-90-1
C7H8BrClFN
240.5
WEGQVRTUTQKDNY-UHFFFAOYSA-N
2-bromo-6-fluorobenzylamine hcl, 2-bromo-6-fluorophenyl methanamine hydrochloride, 2-bromo-6-fluorophenyl methanamine;hydrochloride, 1-2-bromo-6-fluorophenyl methanamine hydrochloride
118798502
(2-bromo-6-fluorofenil)metanamina; clorhidrato
C1=CC(=C(C(=C1)Br)CN)F.Cl
Este artículo no se puede devolver. Vea la política de devoluciones
1214330-90-1
C7H8BrClFN
240.5
WEGQVRTUTQKDNY-UHFFFAOYSA-N
2-bromo-6-fluorobenzylamine hcl, 2-bromo-6-fluorophenyl methanamine hydrochloride, 2-bromo-6-fluorophenyl methanamine;hydrochloride, 1-2-bromo-6-fluorophenyl methanamine hydrochloride
118798502
(2-bromo-6-fluorofenil)metanamina; clorhidrato
C1=CC(=C(C(=C1)Br)CN)F.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Bromo-6-fluorobenzylamine hydrochloride
Cantidad 250 mg
Formula Weight (peso de la fórmula) 240.5
Porcentaje de pureza 96%
Forma física Sólido

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'