missing translation for 'onlineSavingsMsg'
Learn More

Ácido 2-bromo-4,5-dimetoxicinámico, 97 %, Thermo Scientific™

Catalog Number 11442888
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
151539-52-5
C11H11BrO4
287.109
MFCD00017614
MSJCWIFRXCLULX-ONEGZZNKSA-N
2-bromo-4,5-dimethoxycinnamic acid, 6-bromo-3,4-dimethoxycinnamic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, e-3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 2-propenoic acid, 3-2-bromo-4,5-dimethoxyphenyl, e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl-2-propenoic acid
735828
Ácido (E)-3-(2-bromo-4,5-dimetoxifenil)prop-2-enoico
COC1=C(C=C(C(=C1)C=CC(=O)O)Br)OC
Este artículo no se puede devolver. Vea la política de devoluciones
151539-52-5
C11H11BrO4
287.109
MFCD00017614
MSJCWIFRXCLULX-ONEGZZNKSA-N
2-bromo-4,5-dimethoxycinnamic acid, 6-bromo-3,4-dimethoxycinnamic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, e-3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 2-propenoic acid, 3-2-bromo-4,5-dimethoxyphenyl, e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl-2-propenoic acid
735828
Ácido (E)-3-(2-bromo-4,5-dimetoxifenil)prop-2-enoico
COC1=C(C=C(C(=C1)C=CC(=O)O)Br)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Bromo-4,5-dimethoxycinnamic acid
Punto de fusión 253°C to 256°C
Cantidad 5 g
Beilstein 3142451
Formula Weight (peso de la fórmula) 287.12
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'