missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(1-methyl-1H-benzimidazol-5-yl)ethanone hydrobromide, 97%, Thermo Scientific™

Product Code. 10255244
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10255244 1 g Amber Glass Bottle 1g
11320577 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255244 Supplier Thermo Scientific Maybridge Supplier No. CC13655DA
 Restricted Item
Fecha estimada de envoi: 07-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-78-6
Molecular Formula C10H10Br2N2O
Molecular Weight (g/mol) 334.011
MDL Number MFCD09817461
InChI Key LRWGKOQDEBXXPS-UHFFFAOYSA-N
Synonym 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1,3-benzodiazol-5-yl ethanone hydrobromide, c10h9brn2o.hbr, 2-bromo-1-1-methyl-1h-benzoimidazol-5-yl-ethanone.hbr, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one-hydrogen bromide 1/1
PubChem CID 24229460
IUPAC Name 2-bromo-1-(1-methylbenzimidazol-5-yl)ethanone;hydrobromide
SMILES CN1C=NC2=C1C=CC(=C2)C(=O)CBr.Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 334.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(1-methyl-1H-benzimidazol-5-yl)ethanone hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.