Learn More
2-Benciloxi-2-metil-1-propanol, 95 %, Thermo Scientific Chemicals
Catalog Number p-7028614
Click to view available options
Cantidad:
250 mg
1 g
91968-71-7
C11H16O2
180.25
MFCD08459314
FRHJVAJQSPVSKV-UHFFFAOYSA-N
2-benzyloxy-2-methylpropan-1-ol, 2-benzyloxy-2-methyl-1-propanol, 2-methyl-2-phenylmethoxy propan-1-ol
42552903
2-(benzyloxy)-2-methylpropan-1-ol
CC(C)(CO)OCC1=CC=CC=C1
91968-71-7
C11H16O2
180.25
MFCD08459314
FRHJVAJQSPVSKV-UHFFFAOYSA-N
2-benzyloxy-2-methylpropan-1-ol, 2-benzyloxy-2-methyl-1-propanol, 2-methyl-2-phenylmethoxy propan-1-ol
42552903
2-(benzyloxy)-2-methylpropan-1-ol
CC(C)(CO)OCC1=CC=CC=C1
2-Benzyloxy-2-methyl-1-propanol is used as a building block in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El 2-benziloxi-2-metil-1-propanol se utiliza como un compuesto básico en la síntesis orgánica.
Solubilidad
Inmiscible con agua.
Notas
Incompatible con agentes oxidantes fuertes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Benzyloxy-2-methyl-1-propanol
Punto de ebullición 80°C to 85°C (0.1 mmHg)
Cantidad 250 mg
Información de solubilidad Immiscible with water.
Formula Weight (peso de la fórmula) 180.12
Porcentaje de pureza 95%