Learn More
2-Amino-6-iodopurine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-6-iodopurine |
| CAS | 19690-23-4 |
| Molecular Formula | C5H4IN5 |
| Quantity | 10 g |
| Synonym | 6-Iodo-9H-purin-2-amine, NSC 45154 |
| InChI Formula | InChI=1S/C5H4IN5/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H3,7,8,9,10,11) |
| SMILES | IC1=C2N=CN=C2NC(=N)N1 |
| IUPAC Name | 6-iodo-1,3-dihydropurin-2-imine |
| Molecular Weight (g/mol) | 261.02 |
| Formula Weight | 260.9511 |