missing translation for 'onlineSavingsMsg'
Learn More

Hidrobromuro de 2-amino-4,5-dimetiltiazol, 98 %, Thermo Scientific™

Catalog Number p-7025365
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
7170-76-5
C5H9BrN2S
209.105
MFCD00035212
CATCJRCKBGAMKK-UHFFFAOYSA-N
2-amino-4,5-dimethylthiazole hydrobromide, 4,5-dimethyl-1,3-thiazol-2-amine hydrobromide, 4,5-dimethylthiazol-2-amine hydrobromide, unii-nm8hvb696p, 2-thiazolamine, 4,5-dimethyl-, monohydrobromide, nm8hvb696p, 2-thiazolamine, 4,5-dimethyl-, hydrobromide 1:1, dimethyl-1,3-thiazol-2-amine hydrobromide, thiazole, 2-amino-4,5-dimethyl-, hydrobromide, acmc-209ojv
73237
4,5-Dimetil-1,3-tiazol-2-amina; hidrobromuro
CC1=C(SC(=N1)N)C.Br
Este artículo no se puede devolver. Vea la política de devoluciones
7170-76-5
C5H9BrN2S
209.105
MFCD00035212
CATCJRCKBGAMKK-UHFFFAOYSA-N
2-amino-4,5-dimethylthiazole hydrobromide, 4,5-dimethyl-1,3-thiazol-2-amine hydrobromide, 4,5-dimethylthiazol-2-amine hydrobromide, unii-nm8hvb696p, 2-thiazolamine, 4,5-dimethyl-, monohydrobromide, nm8hvb696p, 2-thiazolamine, 4,5-dimethyl-, hydrobromide 1:1, dimethyl-1,3-thiazol-2-amine hydrobromide, thiazole, 2-amino-4,5-dimethyl-, hydrobromide, acmc-209ojv
73237
4,5-Dimetil-1,3-tiazol-2-amina; hidrobromuro
CC1=C(SC(=N1)N)C.Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Amino-4,5-dimethylthiazole hydrobromide
Punto de fusión 287°C to 293°C
Cantidad 5 g
Beilstein 3690705
Formula Weight (peso de la fórmula) 209.11
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'