missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-bromofenil)pirimidina, 95 %, Thermo Scientific™

Catalog Number p-7032661
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
99073-95-7
C10H8BrN3
250.10
MFCD07381905
ZCMWKKYYDONJDU-UHFFFAOYSA-N
4-2-bromophenyl pyrimidin-2-amine, 2-amino-4-2-bromophenyl pyrimidine, 4-2-bromophenyl-2-pyrimidinylamine
53426180
4-(2-bromofenil)pirimidin-2-amina
NC1=NC=CC(=N1)C1=CC=CC=C1Br
Este artículo no se puede devolver. Vea la política de devoluciones
99073-95-7
C10H8BrN3
250.10
MFCD07381905
ZCMWKKYYDONJDU-UHFFFAOYSA-N
4-2-bromophenyl pyrimidin-2-amine, 2-amino-4-2-bromophenyl pyrimidine, 4-2-bromophenyl-2-pyrimidinylamine
53426180
4-(2-bromofenil)pirimidin-2-amina
NC1=NC=CC(=N1)C1=CC=CC=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Amino-4-(2-bromophenyl)pyrimidine
Punto de fusión 167°C to 168°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 250.1
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'