missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 6-Fluoro-DL-triptófano, 97 %
Click to view available options
Cantidad:
100 mg
500 mg
Envase:
Frasco de vidrio
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | 6-Fluoro-DL-tryptophan |
% mín. CAS | 96.0 |
% máx. CAS | 100.0 |
Punto de fusión | 280°C to 285°C |
Color | De beige a blanco |
Espectro infrarrojo | Authentic |
Intervalo de porcentaje de ensayo | 97% |
Envase | Frasco de vidrio |
Cantidad | 100 mg |
Formula Weight (peso de la fórmula) | 222.22 |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'