missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-diyodopiridina, 95 %

Catalog Number p-7029986
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
23597-15-1
C5H4I2N2
345.91
MFCD02663852
QLJWHQFRDOOATP-UHFFFAOYSA-N
350960
3,5-diyodopiridina-2-amina
C1=C(C=NC(=C1I)N)I
Este artículo no se puede devolver. Vea la política de devoluciones
23597-15-1
C5H4I2N2
345.91
MFCD02663852
QLJWHQFRDOOATP-UHFFFAOYSA-N
350960
3,5-diyodopiridina-2-amina
C1=C(C=NC(=C1I)N)I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Amino-3,5-diiodopyridine
Cantidad 250 mg
Sensibilidad Light sensitive
Sinónimo 2-amino-3,5-diiodopyridine, 2-pyridinamine, 3,5-diiodo, acmc-1cip1, 2-pyridinamine,3,5-diiodo, 2-amino-3,5-diiodopyridine 250mg
Formula Weight (peso de la fórmula) 345.91
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'