Learn More
2-(7-methyl-2-oxo-2H-chromen-4-yl)acetic Acid, TRC
Product Code. 30348401
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30348401 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30348401 Supplier Toronto Research Chemicals Supplier No. TRCB499335500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(7-methyl-2-oxo-2H-chromen-4-yl)acetic Acid
CAS 50402-83-0
Molecular Formula C12H10O4
Quantity 500 mg
Synonym (7-Methyl-2-oxo-2H-chromen-4-yl)acetic acid
InChI Formula InChI=1S/C12H10O4/c1-7-2-3-9-8(5-11(13)14)6-12(15)16-10(9)4-7/h2-4,6H,5H2,1H3,(H,13,14)
SMILES CC1=CC2=C(C=C1)C(=CC(=O)O2)CC(=O)O
IUPAC Name 2-(7-methyl-2-oxochromen-4-yl)acetic acid
Molecular Weight (g/mol) 218.2
Formula Weight 218.0579