missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dinitro-9-fluorenona, 97 %, Thermo Scientific Chemicals

Catalog Number 15491338
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
31551-45-8
C13H6N2O5
270.20
MFCD00001153
HDVGAFBXTXDYIB-UHFFFAOYSA-N
2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro
35842
2,7-dinitrofluoren-9-ona
[O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
31551-45-8
C13H6N2O5
270.20
MFCD00001153
HDVGAFBXTXDYIB-UHFFFAOYSA-N
2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro
35842
2,7-dinitrofluoren-9-ona
[O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,7-Dinitro-9-fluorenone
Punto de fusión 292°C to 295°C
Color De amarillo a naranja
Cantidad 100 g
Formula Weight (peso de la fórmula) 270.2
Porcentaje de pureza 97%
Forma física Polvo

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'