missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropil-N,N-dimetilanilina, 96 %, Thermo Scientific Chemicals

Catalog Number p-7029775
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
2909-77-5
C14H23N
205.345
MFCD00144978
ALXIOUGHHXXLKX-UHFFFAOYSA-N
2,6-diisopropyl-n,n-dimethylaniline, n,n-dimethyl-2,6-diisopropylaniline, benzenamine, n,n-dimethyl-2,6-bis 1-methylethyl, 2,6-diisopropyl-n,n-dimethyl-aniline, acmc-20ao8g, 2,6-bis methylethyl phenyl dimethylamine, n,n-dimethyl-2,6-di propan-2-yl aniline, benzenamine,n,n-dimethyl-2,6-bis 1-methylethyl
76198
N,N-dimetil-2,6-di(propan-2-il)anilina
CC(C)C1=C(C(=CC=C1)C(C)C)N(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
2909-77-5
C14H23N
205.345
MFCD00144978
ALXIOUGHHXXLKX-UHFFFAOYSA-N
2,6-diisopropyl-n,n-dimethylaniline, n,n-dimethyl-2,6-diisopropylaniline, benzenamine, n,n-dimethyl-2,6-bis 1-methylethyl, 2,6-diisopropyl-n,n-dimethyl-aniline, acmc-20ao8g, 2,6-bis methylethyl phenyl dimethylamine, n,n-dimethyl-2,6-di propan-2-yl aniline, benzenamine,n,n-dimethyl-2,6-bis 1-methylethyl
76198
N,N-dimetil-2,6-di(propan-2-il)anilina
CC(C)C1=C(C(=CC=C1)C(C)C)N(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,6-Diisopropyl-N,N-dimethylaniline
Densidad 0.873
Punto de ebullición 86°C (2.5 mmHg)
Temperatura de inflamación >110°C (230°F)
Índice de refracción 1.5005
Cantidad 1 g
Número UN UN2810
Formula Weight (peso de la fórmula) 205.34
Porcentaje de pureza 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'