missing translation for 'onlineSavingsMsg'
Learn More

Ácido 2,6-difluorocinámico, 98 %, Thermo Scientific™

Catalog Number p-7025042
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
102082-89-3
C9H6F2O2
184.142
MFCD00010319
JMUOYANNVIFGFN-SNAWJCMRSA-N
2,6-difluorocinnamic acid, trans-2,6-difluorocinnamic acid, e-3-2,6-difluorophenyl acrylic acid, 3-2,6-difluorophenyl acrylic acid, 2e-3-2,6-difluorophenyl prop-2-enoic acid, 2e-3-2,6-difluorophenyl acrylic acid, 3-2,6-difluoro-phenyl-acrylic acid, e-3-2,6-difluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2,6-difluorophenyl-, 2e, rarechem bk hw 0116
5372267
Ácido (E)-3-(2,6-difluorofenil)prop-2-enoico
C1=CC(=C(C(=C1)F)C=CC(=O)O)F
Este artículo no se puede devolver. Vea la política de devoluciones
102082-89-3
C9H6F2O2
184.142
MFCD00010319
JMUOYANNVIFGFN-SNAWJCMRSA-N
2,6-difluorocinnamic acid, trans-2,6-difluorocinnamic acid, e-3-2,6-difluorophenyl acrylic acid, 3-2,6-difluorophenyl acrylic acid, 2e-3-2,6-difluorophenyl prop-2-enoic acid, 2e-3-2,6-difluorophenyl acrylic acid, 3-2,6-difluoro-phenyl-acrylic acid, e-3-2,6-difluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2,6-difluorophenyl-, 2e, rarechem bk hw 0116
5372267
Ácido (E)-3-(2,6-difluorofenil)prop-2-enoico
C1=CC(=C(C(=C1)F)C=CC(=O)O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,6-Difluorocinnamic acid
Punto de fusión 124°C to 125°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 184.14
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'