Learn More
2,6-Difluoro-4-methoxyaniline, 97%, Thermo Scientific™
Product Code. 11322709
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322709 1 g 1g
11332709 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322709 Supplier Thermo Scientific Alfa Aesar Supplier No. H26728.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,6-Difluoro-4-methoxyaniline is used as primary and secondary intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 151414-47-0
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.136
MDL Number MFCD04115922
InChI Key HQBURSNJKAITGQ-UHFFFAOYSA-N
Synonym 4-amino-3,5-difluoroanisole, 2,6-difluoro-4-methoxyphenylamine, benzenamine, 2,6-difluoro-4-methoxy, 2,6-difluoro-4-methoxy-benzenamine, 2,6-difluoro-4-methoxy-phenylamine, intermediates-zcf02157, acmc-209z8h, 4-methoxy-2,6-difluoroaniline, 2,6-difluoro-4-methoxybenzenamine, benzenamine,2,6-difluoro-4-methoxy
PubChem CID 5185763
IUPAC Name 2,6-difluoro-4-methoxyaniline
SMILES COC1=CC(=C(C(=C1)F)N)F
Specifications
Melting Point 38°C to 40°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-4-methoxyaniline