Learn More
2,5-Pyridinedicarboxylic Acid, TRC
Product Code. 30413634
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413634 50 g 50g
30410346 25 g -
30410855 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30413634 Supplier Toronto Research Chemicals Supplier No. TRCP99164050G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-26-5
Molecular Formula C7 H5 N O4
Molecular Weight (g/mol) 167.12
Synonym Pyridine-2,5-dicarboxylic Acid, 2,5-Pyridinedicarboxylic acid, 2,5-Dicarboxypyridine, Isocinchomeronic acid, NSC 177
IUPAC Name pyridine-2,5-dicarboxylic acid
SMILES OC(=O)c1ccc(nc1)C(=O)O
Specifications
Quantity 50 g
InChI Formula InChI=1S/C7H5NO4/c9-6(10)4-1-2-5(7(11)12)8-3-4/h1-3H,(H,9,10)(H,11,12)
Formula Weight 167.0219
Chemical Name or Material Isocinchomeronic Acid