Learn More
2',5'-Difluoroacetophenone, TRC
Especificaciones
Especificaciones
| Chemical Name or Material | 2',5'-Difluoroacetophenone |
| CAS | 1979-36-8 |
| Molecular Formula | C8H6F2O |
| Quantity | 1 g |
| Synonym | 1-(2,5-Difluorophenyl)ethanone, 2,5-Difluorophenyl Methyl Ketone |
| InChI Formula | InChI=1S/C8H6F2O/c1-5(11)7-4-6(9)2-3-8(7)10/h2-4H,1H3 |
| SMILES | CC(=O)c1cc(F)ccc1F |
| IUPAC Name | 1-(2,5-difluorophenyl)ethanone |
| Molecular Weight (g/mol) | 156.13 |
| Formula Weight | 156.0387 |