missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromoaniline, 97%

Product Code. 11419416
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419416 100 g 100g
11499406 5 g 5g
11409416 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419416 Supplier Thermo Scientific Alfa Aesar Supplier No. A19114.22
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy

CAS: 3638-73-1 | C6H5Br2N | 250.921 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3638-73-1
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00007636
InChI Key WRTAZRGRFBCKBU-UHFFFAOYSA-N
Synonym 2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline
PubChem CID 77198
IUPAC Name 2,5-dibromoaniline
SMILES C1=CC(=C(C=C1Br)N)Br

Specifications

Melting Point 51°C to 56°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 100 g
UN Number UN2811
Beilstein 1817548
Formula Weight 250.93
Percent Purity 98%
Chemical Name or Material 2,5-Dibromoaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.