missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorofenil)-2-metilmorfolina, 99 %, Thermo Scientific Chemicals

Catalog Number p-7033831
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
109461-46-3
C11H14FNO
195.237
MFCD08061112
IHMLZEWIFPIGKG-UHFFFAOYSA-N
2-4-fluorophenyl-2-methylmorpholine, 2-4-fluorophenyl-2-methyl-morpholine, acmc-1c5kc, morpholine,2-4-fluorophenyl-2-methyl
3066047
2-(4-fluorofenil)-2-metilmorfolina
CC1(CNCCO1)C2=CC=C(C=C2)F
Este artículo no se puede devolver. Vea la política de devoluciones
109461-46-3
C11H14FNO
195.237
MFCD08061112
IHMLZEWIFPIGKG-UHFFFAOYSA-N
2-4-fluorophenyl-2-methylmorpholine, 2-4-fluorophenyl-2-methyl-morpholine, acmc-1c5kc, morpholine,2-4-fluorophenyl-2-methyl
3066047
2-(4-fluorofenil)-2-metilmorfolina
CC1(CNCCO1)C2=CC=C(C=C2)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-(4-Fluorophenyl)-2-methylmorpholine
Cantidad 250 mg
Formula Weight (peso de la fórmula) 195.24
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'