missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%

Product Code. 15441156
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441156 1 g 1g
15461156 250 mg 250mg
15451156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441156 Supplier Thermo Scientific Alfa Aesar Supplier No. H54410.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28485-18-9 | C4H3ClN2O4S | 210.59 g/mol

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28485-18-9
Molecular Formula C4H3ClN2O4S
Molecular Weight (g/mol) 210.59
MDL Number MFCD06655485
InChI Key FQHNJCWWBKYCJW-UHFFFAOYSA-N
Synonym 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride, 1,2,3,4-tetrahydro-2,4-dioxo-5-pyrimidinesulfonyl chloride, 5-pyrimidinesulfonylchloride, 1,2,3,4-tetrahydro-2,4-dioxo, uracil-5-sulphonyl chloride, 5-chlorosulfonyl-1,3-dihydropyrimidine-2,4-dione, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinesulfonyl chloride, 2,4-dioxo-1,2,3,4-tetrahydro-pyrimidine-5-sulfonyl chloride
PubChem CID 234620
SMILES ClS(=O)(=O)C1=CNC(=O)NC1=O

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 2,4-dioxo-1H-pyrimidine-5-sulfonyl chloride
Formula Weight 209.95
Percent Purity 97%
Chemical Name or Material 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.