missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimetilfenilurea, 98 %, Thermo Scientific™

Catalog Number p-7028381
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
2990-02-5
C9H12N2O
164.21
MFCD00025409
GKEHHLJOYZDXMI-UHFFFAOYSA-N
2,4-dimethylphenyl urea, 1-2,4-dimethylphenyl urea, n-2,4-dimethylphenyl urea, 2,4-xylylurea, 5-chlorobenzoic acid, acmc-20aoc5, urea,n-2,4-dimethylphenyl, amino-n-2,4-dimethylphenyl amide
853638
(2,4-dimetilfenil)urea
CC1=CC(=C(C=C1)NC(=O)N)C
Este artículo no se puede devolver. Vea la política de devoluciones
2990-02-5
C9H12N2O
164.21
MFCD00025409
GKEHHLJOYZDXMI-UHFFFAOYSA-N
2,4-dimethylphenyl urea, 1-2,4-dimethylphenyl urea, n-2,4-dimethylphenyl urea, 2,4-xylylurea, 5-chlorobenzoic acid, acmc-20aoc5, urea,n-2,4-dimethylphenyl, amino-n-2,4-dimethylphenyl amide
853638
(2,4-dimetilfenil)urea
CC1=CC(=C(C=C1)NC(=O)N)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,4-Dimethylphenylurea
Punto de fusión 203°C to 204°C (decomposition)
Cantidad 5 g
Formula Weight (peso de la fórmula) 164.2
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'