missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimetil-3-heptanol, eritro + treo, 98 %, Thermo Scientific™

Catalog Number 11440701
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
19549-72-5
C9H20O
144.258
MFCD00027148
PSCLFHNWSAXNJL-UHFFFAOYSA-N
2,4-dimethyl-3-heptanol, 3-heptanol, 2,4-dimethyl, acmc-20aocf, 3-heptanol,4-dimethyl, 3-heptanol,2,4-dimethyl, 2,4-dimethyl-3-heptanol, threo + erythro, 2,4-dimethyl-3-heptanol, erythro + threo
140545
2,4-Dimetilheptan-3-ol
CCCC(C)C(C(C)C)O
Este artículo no se puede devolver. Vea la política de devoluciones
19549-72-5
C9H20O
144.258
MFCD00027148
PSCLFHNWSAXNJL-UHFFFAOYSA-N
2,4-dimethyl-3-heptanol, 3-heptanol, 2,4-dimethyl, acmc-20aocf, 3-heptanol,4-dimethyl, 3-heptanol,2,4-dimethyl, 2,4-dimethyl-3-heptanol, threo + erythro, 2,4-dimethyl-3-heptanol, erythro + threo
140545
2,4-Dimetilheptan-3-ol
CCCC(C)C(C(C)C)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,4-Dimethyl-3-heptanol
Nombre de nota erythro + threo
Cantidad 25 g
Formula Weight (peso de la fórmula) 144.25
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'