Learn More
2,4-Dichlorobenzotrifluoride, TRC
Product Code. 30011644
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011644 10 g 10g
30007069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011644 Supplier Toronto Research Chemicals Supplier No. TRCD43197010G
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 320-60-5
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 215
Synonym 1,3-Dichloro-4-(trifluoromethyl)benzene, 2,4-Dichloro(trifluoromethyl)benzene, 2,4-Dichloro-1-(trifluoromethyl)benzene, 2,4-Dichloro-α,α,α-trifluorotoluene, 2,4-Dichloro-α,α,α-trifluorotoluene, 2,4-Dichloro-1-(trifluoromethyl)benzene
SMILES FC(F)(F)c1ccc(Cl)cc1Cl
Specifications
Quantity 10 g
Chemical Name or Material 2,4-Dichlorobenzotrifluoride