missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dicloro-5-nitrobenzotrifluoruro, 97 %, Thermo Scientific™

Catalog Number p-7023058
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
400-70-4
C7H2Cl2F3NO2
259.993
MFCD00007073
VLVNHMVSVDVAOA-UHFFFAOYSA-N
2,4-dichloro-5-nitrobenzotrifluoride, 1,5-dichloro-2-nitro-4-trifluoromethyl benzene, 2,4-dichloro-3,5-nitrobenzotrifluoride, benzene, 1,5-dichloro-2-nitro-4-trifluoromethyl, 2,4-dichloro-5-trifluoromethyl-1-nitrobenzene, pubchem4862, 2,4-dichloro-5-nitro benzotrifluoride, 3-trifluoromethyl-4,6-dichloronitrobenzene, 2,4-dichloro-5-trifluoromethyl nitrobenzene
621179
1,5-dicloro-2-nitro-4-(trifluorometil)benceno
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
400-70-4
C7H2Cl2F3NO2
259.993
MFCD00007073
VLVNHMVSVDVAOA-UHFFFAOYSA-N
2,4-dichloro-5-nitrobenzotrifluoride, 1,5-dichloro-2-nitro-4-trifluoromethyl benzene, 2,4-dichloro-3,5-nitrobenzotrifluoride, benzene, 1,5-dichloro-2-nitro-4-trifluoromethyl, 2,4-dichloro-5-trifluoromethyl-1-nitrobenzene, pubchem4862, 2,4-dichloro-5-nitro benzotrifluoride, 3-trifluoromethyl-4,6-dichloronitrobenzene, 2,4-dichloro-5-trifluoromethyl nitrobenzene
621179
1,5-dicloro-2-nitro-4-(trifluorometil)benceno
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,4-Dichloro-5-nitrobenzotrifluoride
Punto de fusión 53°C to 57°C
Temperatura de inflamación >110°C (230°F)
Cantidad 5 g
Beilstein 2467459
Formula Weight (peso de la fórmula) 260
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'