Learn More
Alfa Aesar™ 2,4-Dichloro-5-(chloromethyl)pyrimidine, 95%
Marca:  Alfa Aesar™ H33223.03
Detalles adicionales : CAS : 7627-38-5 Peso : 0.00100kg
Cantidad. 1g
 Ver más versiones de este producto
Código de producto. 15496027
  • 205.10€ / 1 g
Fecha de envío estimada
del proveedor 07-10-2019
Añadir a la cesta
Descripción
Descripción
Applications
Various pyrimidines having an olefinic side chain were also synthesized via the Wittig reagents derived from chloromethylpyrimidines.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Especificaciones
7627-38-5
197.443
VLOODHWTRPYFIS-UHFFFAOYSA-N
13568279
C1=C(C(=NC(=N1)Cl)Cl)CCl
C5H3Cl3N2
MFCD10697639
2,4-dichloro-5-chloromethyl pyrimidine, pyrimidine, 2,4-dichloro-5-chloromethyl, dichlorochloromethylpyrimidine, 2,4-dichlor-5-chlormethylpyrimidin, 2,4-dichloro-5-choromethyl-pyrimidine, 2,4-dichloro-5-chloromethyl-pyrimidine
2,4-dichloro-5-(chloromethyl)pyrimidine
Especificaciones
1g
White to Cream
7627-38-5
MFCD10697639
VLOODHWTRPYFIS-UHFFFAOYSA-N
2,4-dichloro-5-(chloromethyl)pyrimidine
13568279
Solid
2,4-Dichloro-5-(chloromethyl)pyrimidine
63°C to 65°C
C5H3Cl3N2
2,4-dichloro-5-chloromethyl pyrimidine, pyrimidine, 2,4-dichloro-5-chloromethyl, dichlorochloromethylpyrimidine, 2,4-dichlor-5-chlormethylpyrimidin, 2,4-dichloro-5-choromethyl-pyrimidine, 2,4-dichloro-5-chloromethyl-pyrimidine
C1=C(C(=NC(=N1)Cl)Cl)CCl
197.443
197.5
95%
Videos
SDS Documentos