missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triclorotiofenol, 97 %, Thermo Scientific Chemicals

Catalog Number p-7043888
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
3773-14-6
C6H3Cl3S
213.5
MFCD00014447
JARIALSGFXECCH-UHFFFAOYSA-N
2,4,5-trichlorothiophenol, renacit ii, benzenethiol, 2,4,5-trichloro, 2,4,5-trichlorobenzene-1-thiol, acmc-209iv9, 2,4,5-trichloro-thiophenol, 2,4,5-trichlorophenyl mercaptan, 2,4,5-tris chloranyl benzenethiol, n-memet-n-trimetsilylmet benzylamine
19597
2,4,5-triclorobencenotiol
C1=C(C(=CC(=C1Cl)Cl)Cl)S
Este artículo no se puede devolver. Vea la política de devoluciones
3773-14-6
C6H3Cl3S
213.5
MFCD00014447
JARIALSGFXECCH-UHFFFAOYSA-N
2,4,5-trichlorothiophenol, renacit ii, benzenethiol, 2,4,5-trichloro, 2,4,5-trichlorobenzene-1-thiol, acmc-209iv9, 2,4,5-trichloro-thiophenol, 2,4,5-trichlorophenyl mercaptan, 2,4,5-tris chloranyl benzenethiol, n-memet-n-trimetsilylmet benzylamine
19597
2,4,5-triclorobencenotiol
C1=C(C(=CC(=C1Cl)Cl)Cl)S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,4,5-Trichlorothiophenol
Punto de fusión 112°C to 118°C
Punto de ebullición 153°C to 154°C (15 mmHg)
Olor Unpleasant
Cantidad 1 g
Número UN UN3335
Beilstein 509727
Sensibilidad Air Sensitive
Formula Weight (peso de la fórmula) 213.51
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'