missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimetilpentano, 97 %, Thermo Scientific Chemicals

Catalog Number p-7058973
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
565-59-3
C7H16
100.21
MFCD00009337
WGECXQBGLLYSFP-UHFFFAOYNA-N
pentane, 2,3-dimethyl, 3,4-dimethylpentane, acmc-1an9i, 3-01-00-00445 beilstein handbook reference, 2,3-dimethylpentane, 2,3-dimethylpentane, analytical standard
11260
2,3-dimetilpentano
CCC(C)C(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
565-59-3
C7H16
100.21
MFCD00009337
WGECXQBGLLYSFP-UHFFFAOYNA-N
pentane, 2,3-dimethyl, 3,4-dimethylpentane, acmc-1an9i, 3-01-00-00445 beilstein handbook reference, 2,3-dimethylpentane, 2,3-dimethylpentane, analytical standard
11260
2,3-dimetilpentano
CCC(C)C(C)C

It is used in the liquid-phase oxidation of 2,4-dimethylpentane and in a study on the preparation and the physical constants of a number of alkanes and cycloalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,3-Dimethylpentane
Densidad 0.690g/mL
Punto de ebullición 89°C to 90°C
Temperatura de inflamación −6°C (21°F)
Índice de refracción 1.3920
Cantidad 1 g
Número UN UN1206
Formula Weight (peso de la fórmula) 100.21
Porcentaje de pureza 97%
Forma física Líquido

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'