missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-3-heptanol, 94%, Thermo Scientific™

Product Code. 11454467
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454467 1 g 1g
11464467 5 g 5g
11474467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454467 Supplier Thermo Scientific Alfa Aesar Supplier No. B20480.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19549-71-4
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
MDL Number MFCD00046605
InChI Key JIEGVNXCNNWVPH-UHFFFAOYNA-N
PubChem CID 140544
IUPAC Name 2,3-dimethylheptan-3-ol
SMILES CCCCC(C)(O)C(C)C

Specifications

Density 0.83
Quantity 1 g
Synonym 2,3-dimethyl-3-heptanol, 3-heptanol, 2,3-dimethyl, 3-heptanol,2,3-dimethyl, acmc-1bs02
Formula Weight 144.25
Percent Purity 94%
Chemical Name or Material 2,3-Dimethyl-3-heptanol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.