missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-metilbenzamida, +97 %, Thermo Scientific™

Catalog Number p-7027475
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
261763-35-3
C8H7F2NO
171.15
MFCD01631633
JWPKXGWPXBRCLL-UHFFFAOYSA-N
acmc-1cq0b, 2,3-difluoro-p-toluamide, 2,3-difluoro-4-methyl-benzamide, benzamide,2,3-difluoro-4-methyl, benzamide, 2,3-difluoro-4-methyl, 2,3-bis fluoranyl-4-methyl-benzamide
2774139
2,3-difluoro-4-metilbenzamida
CC1=CC=C(C(N)=O)C(F)=C1F
Este artículo no se puede devolver. Vea la política de devoluciones
261763-35-3
C8H7F2NO
171.15
MFCD01631633
JWPKXGWPXBRCLL-UHFFFAOYSA-N
acmc-1cq0b, 2,3-difluoro-p-toluamide, 2,3-difluoro-4-methyl-benzamide, benzamide,2,3-difluoro-4-methyl, benzamide, 2,3-difluoro-4-methyl, 2,3-bis fluoranyl-4-methyl-benzamide
2774139
2,3-difluoro-4-metilbenzamida
CC1=CC=C(C(N)=O)C(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,3-Difluoro-4-methylbenzamide
Punto de fusión 158°C to 162°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 171.15
Porcentaje de pureza ≥97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'